Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 117in
Catalog No. 502765857
Encompass_Preferred
The Krueger 16N497 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window to allow visual monitoring of oil level and a bronze body for corrosion resistance. This model has a 3/4" NPT thread and is adjustable from 4" to 12" in length. It is made in the USA and is rated for pressures up to 1000 psi at temperatures ranging from -40°F to 250°F. The gauge has a dual scale that reads in inches on one side and centimeters on the other.
Catalog No. 50-276-5857 Supplier Grainger Supplier No. 16N497
The Krueger 16N497 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window to allow visual monitoring of oil level and a bronze body for corrosion resistance. This model has a 3/4" NPT thread and is adjustable from 4" to 12" in length. It is made in the USA and is rated for pressures up to 1000 psi at temperatures ranging from -40°F to 250°F. The gauge has a dual scale that reads in inches on one side and centimeters on the other.