Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 99in
Catalog No. 502765953
Encompass_Preferred
The Krueger 16N480 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a polypropylene indicator head. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. It has a stem length of 48 inches and indicates liquid levels on a clearly marked scale in inches and centimeters. The gauge can be mounted vertically or horizontally through a 3/4 inch NPT opening. This rugged and durable gauge provides accurate liquid level indication for industrial applications.
Catalog No. 50-276-5953 Supplier Grainger Supplier No. 16N480
The Krueger 16N480 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a polypropylene indicator head. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. It has a stem length of 48 inches and indicates liquid levels on a clearly marked scale in inches and centimeters. The gauge can be mounted vertically or horizontally through a 3/4 inch NPT opening. This rugged and durable gauge provides accurate liquid level indication for industrial applications.