Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 109in
Catalog No. 502765954
Encompass_Preferred
The Krueger 16N489 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green, yellow, and red zones for quick visual indication of fluid level. The gauge body is made of steel and mounts into a standard 1/2" NPT threaded port. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The gauge window measures 2-1/4" wide x 4-1/2" high. This rugged and economical gauge provides an easy way to monitor fluid levels.
Catalog No. 50-276-5954 Supplier Grainger Supplier No. 16N489
The Krueger 16N489 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green, yellow, and red zones for quick visual indication of fluid level. The gauge body is made of steel and mounts into a standard 1/2" NPT threaded port. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The gauge window measures 2-1/4" wide x 4-1/2" high. This rugged and economical gauge provides an easy way to monitor fluid levels.