Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 93in
Catalog No. 502765955
Encompass_Preferred
The Krueger Level Gauge For 93 in Container is a float-operated mechanical level gauge designed for use with 93 inch tall industrial containers. It features a hermetically sealed float for reliability, 316 stainless steel wetted parts for corrosion resistance, and a universal mounting bracket that allows top or side mounting. The gauge indicates liquid level on an easy-to-read dial with increments from 0 to 100 percent. This heavy duty industrial gauge helps monitor liquid levels in large containers up to 93 inches tall.
Catalog No. 50-276-5955 Supplier Grainger Supplier No. 16N475
The Krueger Level Gauge For 93 in Container is a float-operated mechanical level gauge designed for use with 93 inch tall industrial containers. It features a hermetically sealed float for reliability, 316 stainless steel wetted parts for corrosion resistance, and a universal mounting bracket that allows top or side mounting. The gauge indicates liquid level on an easy-to-read dial with increments from 0 to 100 percent. This heavy duty industrial gauge helps monitor liquid levels in large containers up to 93 inches tall.