Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 116in
Catalog No. 502765990
Encompass_Preferred
The Krueger Level Gauge For 89 in Container is a mechanical level gauge designed for 89 inch high containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and mounts to the top of the container with a 3/4 inch NPT connection. This heavy duty mechanical gauge provides accurate level indication without any power requirements. Made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-5990 Supplier Grainger Supplier No. 16N496
The Krueger Level Gauge For 89 in Container is a mechanical level gauge designed for 89 inch high containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and mounts to the top of the container with a 3/4 inch NPT connection. This heavy duty mechanical gauge provides accurate level indication without any power requirements. Made in the USA by Krueger Sentry Gauge.