Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 17in
Catalog No. 502764071
Encompass_Preferred
The Krueger 16N409 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a clear acrylic lens for viewing fluid level. The gauge has a dual scale that reads in inches and centimeters with a maximum reading of 8 inches or 20 centimeters. It has a 1/2 inch NPT thread connection and includes a locknut for securing it to the reservoir. The gauge can withstand operating temperatures from -40 to 250 degrees Fahrenheit. Overall, this is a sturdy, dual-scale liquid level gauge suitable for hydraulic fluid reservoirs.
Catalog No. 50-276-4071 Supplier Grainger Supplier No. 16N409
The Krueger 16N409 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a clear acrylic lens for viewing fluid level. The gauge has a dual scale that reads in inches and centimeters with a maximum reading of 8 inches or 20 centimeters. It has a 1/2 inch NPT thread connection and includes a locknut for securing it to the reservoir. The gauge can withstand operating temperatures from -40 to 250 degrees Fahrenheit. Overall, this is a sturdy, dual-scale liquid level gauge suitable for hydraulic fluid reservoirs.