Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 100in
Catalog No. 502766022
Encompass_Preferred
The Krueger 16N481 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and process tanks to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and an aluminum case. It is made in the USA and is 36 inches long. The maximum pressure rating is 30 psi at 406°F. This level gauge has dual colored flags to indicate level - red flags for low level and green flags for high level. It can be mounted horizontally or vertically. Overall, this is a heavy duty magnetic level gauge for industrial applications requiring liquid level indication.
Catalog No. 50-276-6022 Supplier Grainger Supplier No. 16N481
The Krueger 16N481 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and process tanks to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and an aluminum case. It is made in the USA and is 36 inches long. The maximum pressure rating is 30 psi at 406°F. This level gauge has dual colored flags to indicate level - red flags for low level and green flags for high level. It can be mounted horizontally or vertically. Overall, this is a heavy duty magnetic level gauge for industrial applications requiring liquid level indication.