Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 35in
Catalog No. 502766084
Encompass_Preferred
The Krueger 16N425 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has 1/4 inch NPT process connections and is compatible with water-based liquids including diesel fuel, solvents, water, and antifreeze. It can be mounted vertically or horizontally. This American made gauge comes with a 5 year warranty from Krueger.
Catalog No. 50-276-6084 Supplier Grainger Supplier No. 16N425
The Krueger 16N425 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has 1/4 inch NPT process connections and is compatible with water-based liquids including diesel fuel, solvents, water, and antifreeze. It can be mounted vertically or horizontally. This American made gauge comes with a 5 year warranty from Krueger.