Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 118in
Catalog No. 502766090
Encompass_Preferred
The Krueger 16N498 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed glass tube for viewing fluid level. It has dual mounting thread options of 1 inch NPT and 1 inch BSPP with a maximum pressure rating of 100 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has an accuracy of 3% full scale and includes isolation valves for ease of maintenance. This industrial liquid level gauge provides a clear visual indication of tank level for operations monitoring and control.
Catalog No. 50-276-6090 Supplier Grainger Supplier No. 16N498
The Krueger 16N498 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed glass tube for viewing fluid level. It has dual mounting thread options of 1 inch NPT and 1 inch BSPP with a maximum pressure rating of 100 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has an accuracy of 3% full scale and includes isolation valves for ease of maintenance. This industrial liquid level gauge provides a clear visual indication of tank level for operations monitoring and control.