Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 114in
Catalog No. 502768432
Encompass_Preferred
The Krueger Level Gauge For 114 in Container is a float-operated mechanical level gauge designed for use with 114 inch diameter tanks. It features a hermetically sealed gauge head and float for use with water, diesel fuel, solvents, oil and other non-corrosive liquids. The polypropylene float has a specific gravity of 0.9 and the stainless steel operating lever ensures smooth operation. This mechanical tank level gauge allows visual monitoring of liquid level at a glance. It is easy to install and requires no electricity. The Krueger model # 16N494 level gauge is ideal for applications where visual liquid level indication is needed on large tanks up to 114 inches in diameter.
Catalog No. 50-276-8432 Supplier Grainger Supplier No. 16N494
The Krueger Level Gauge For 114 in Container is a float-operated mechanical level gauge designed for use with 114 inch diameter tanks. It features a hermetically sealed gauge head and float for use with water, diesel fuel, solvents, oil and other non-corrosive liquids. The polypropylene float has a specific gravity of 0.9 and the stainless steel operating lever ensures smooth operation. This mechanical tank level gauge allows visual monitoring of liquid level at a glance. It is easy to install and requires no electricity. The Krueger model # 16N494 level gauge is ideal for applications where visual liquid level indication is needed on large tanks up to 114 inches in diameter.