Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 61in
Catalog No. 502768502
Encompass_Preferred
The Krueger 61-inch Level Gauge is a liquid level indicator designed for 61-inch containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The scale is easy to read and shows level in inches and centimeters. This economical gauge helps monitor liquid levels at a glance. It has a maximum temperature rating of 180°F and fits standard 2-inch fittings. The Krueger model 16N446 level gauge is ideal for monitoring liquid levels in 61-inch tanks or containers.
Catalog No. 50-276-8502 Supplier Grainger Supplier No. 16N446
The Krueger 61-inch Level Gauge is a liquid level indicator designed for 61-inch containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The scale is easy to read and shows level in inches and centimeters. This economical gauge helps monitor liquid levels at a glance. It has a maximum temperature rating of 180°F and fits standard 2-inch fittings. The Krueger model 16N446 level gauge is ideal for monitoring liquid levels in 61-inch tanks or containers.