Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 62in
Catalog No. 502768503
Encompass_Preferred
The Krueger 16N447 is a liquid level gauge designed for industrial applications. It features a stainless steel float and polypropylene body, with a maximum temperature rating of 250°F. This gauge has a 4 inch scale and indicates liquid level visually. It can be mounted to the top or side of a tank through either the 3/4" NPT bottom or back 1/2" NPT connections. The 16N447 has accuracy within 1/8" and a 10 psi maximum pressure rating. This economical gauge is ideal for monitoring liquid levels in compatible tanks and vessels.
Catalog No. 50-276-8503 Supplier Grainger Supplier No. 16N447
The Krueger 16N447 is a liquid level gauge designed for industrial applications. It features a stainless steel float and polypropylene body, with a maximum temperature rating of 250°F. This gauge has a 4 inch scale and indicates liquid level visually. It can be mounted to the top or side of a tank through either the 3/4" NPT bottom or back 1/2" NPT connections. The 16N447 has accuracy within 1/8" and a 10 psi maximum pressure rating. This economical gauge is ideal for monitoring liquid levels in compatible tanks and vessels.