Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 66in
Catalog No. 502767288
Encompass_Preferred
The Krueger Level Gauge For 66 in Container is a float-operated mechanical level gauge designed for use with 66 inch high containers. It features a hermetically sealed reed switch that activates indicator lights to provide a visual display of liquid level. This stainless steel gauge has an adjustable stem length from 34 to 64 inches and provides high, mid and low level indication. It can handle temperatures up to 250°F and pressures up to 100 PSI. The Krueger level gauge offers durable construction and easy installation for industrial liquid level measurement.
Catalog No. 50-276-7288 Supplier Grainger Supplier No. 16N450
The Krueger Level Gauge For 66 in Container is a float-operated mechanical level gauge designed for use with 66 inch high containers. It features a hermetically sealed reed switch that activates indicator lights to provide a visual display of liquid level. This stainless steel gauge has an adjustable stem length from 34 to 64 inches and provides high, mid and low level indication. It can handle temperatures up to 250°F and pressures up to 100 PSI. The Krueger level gauge offers durable construction and easy installation for industrial liquid level measurement.