Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 113in
Catalog No. 502767429
Encompass_Preferred
The Krueger 16N493 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window to allow visual monitoring of oil level and a bronze body for corrosion resistance. This model has a 3/4" NPT thread and is adjustable from 4" to 12" in length. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The Krueger 16N493 provides a simple way to monitor fluid levels in industrial equipment.
Catalog No. 50-276-7429 Supplier Grainger Supplier No. 16N493
The Krueger 16N493 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window to allow visual monitoring of oil level and a bronze body for corrosion resistance. This model has a 3/4" NPT thread and is adjustable from 4" to 12" in length. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The Krueger 16N493 provides a simple way to monitor fluid levels in industrial equipment.