Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 102in
Catalog No. 502768340
Encompass_Preferred
The Krueger 16N483 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate sight glass. It has brass 1/4 inch NPT process connections and is rated for pressures up to 250 PSI at temperatures up to 250°F. The gauge allows direct reading of liquid level and can be mounted vertically or horizontally. This is an economical mechanical gauge for basic liquid level indication.
Catalog No. 50-276-8340 Supplier Grainger Supplier No. 16N483
The Krueger 16N483 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate sight glass. It has brass 1/4 inch NPT process connections and is rated for pressures up to 250 PSI at temperatures up to 250°F. The gauge allows direct reading of liquid level and can be mounted vertically or horizontally. This is an economical mechanical gauge for basic liquid level indication.