Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 94in
Catalog No. 502768385
Encompass_Preferred
The Krueger 16N476 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The stem lengths available range from 7" to 78". This rugged and economical gauge is ideal for use with water, oil and other liquids in tanks, totes and day tanks. With proper installation, it can provide reliable liquid level indication.
Catalog No. 50-276-8385 Supplier Grainger Supplier No. 16N476
The Krueger 16N476 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The stem lengths available range from 7" to 78". This rugged and economical gauge is ideal for use with water, oil and other liquids in tanks, totes and day tanks. With proper installation, it can provide reliable liquid level indication.