Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 101in
Catalog No. 502768641
Encompass_Preferred
The Krueger 16N482 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has 1/4 inch NPT process connections and is compatible with water-based liquids including diesel fuel, solvents, water, and antifreeze. This is an economical mechanical dial gauge for basic liquid level indication.
Catalog No. 50-276-8641 Supplier Grainger Supplier No. 16N482
The Krueger 16N482 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has 1/4 inch NPT process connections and is compatible with water-based liquids including diesel fuel, solvents, water, and antifreeze. This is an economical mechanical dial gauge for basic liquid level indication.