Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 95in
Catalog No. 502768642
Encompass_Preferred
The Krueger 16N477 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual indication of fluid level. The gauge body is made of steel and mounts into a standard 1/2" NPT threaded port. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The gauge allows the user to easily check fluid levels without shutting down equipment. With its durable construction, it is well-suited for industrial environments.
Catalog No. 50-276-8642 Supplier Grainger Supplier No. 16N477
The Krueger 16N477 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual indication of fluid level. The gauge body is made of steel and mounts into a standard 1/2" NPT threaded port. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The gauge allows the user to easily check fluid levels without shutting down equipment. With its durable construction, it is well-suited for industrial environments.