Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 34in
Catalog No. 502763707
Encompass_Preferred
The Krueger 16N424 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2" diameter. The gauge meets ASME B31.1 and B31.3 requirements. With a maximum pressure rating of 30 psi @ 100°F, this level gauge provides an economical solution for monitoring liquid levels in low pressure applications. The 10 inch scale allows operators to visually inspect and monitor tank level changes.
Catalog No. 50-276-3707 Supplier Grainger Supplier No. 16N424
The Krueger 16N424 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2" diameter. The gauge meets ASME B31.1 and B31.3 requirements. With a maximum pressure rating of 30 psi @ 100°F, this level gauge provides an economical solution for monitoring liquid levels in low pressure applications. The 10 inch scale allows operators to visually inspect and monitor tank level changes.