Learn More
ATCC G-361; Melanoma; Human (Homo sapiens)
Catalog No. 502383378
Encompass
G-361 is a cell that has an epithelial morphology that was isolated from the skin of a White, 31-year-old, male with malignant melanoma. This cell line was deposited by PT Peebles and can be used in toxicology research.
Catalog No. 50-238-3378 Supplier ATCC Supplier No. CRL1424
G-361 is a cell that has an epithelial morphology that was isolated from the skin of a White, 31-year-old, male with malignant melanoma. This cell line was deposited by PT Peebles and can be used in toxicology research.