Learn More
Aviva Systems Biology Corp G3BP1 Antibody - N-terminal region (ARP37714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against G3BP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human G3BP1. Alternate names: G3BP, HDH-VIII, MGC111040ApplicationWB, IP, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56405