Learn More
ABclonal Technology G3BP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14836500UL
Encompass_Preferred
This gene encodes one of the DNA-unwinding enzymes which prefers partially unwound 3-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion. This enzyme is a member of the heterogeneous nuclear RNA-binding proteins and is also an element of the Ras signal transduction pathway. It binds specifically to the Ras-GTPase-activating protein by associating with its SH3 domain. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Catalog No. 50-001-05079