Learn More
ATCC G-401; Kidney Tumor; Human (Homo sapiens)
Catalog No. 502384363
Encompass
The G-401 [G401] is epithelial-like and isolated from the kidney of a White, 3-month, male with a rhabdoid tumor. This cell line was deposited by PT Peebles.
Catalog No. 50-238-4363 Supplier ATCC Supplier No. CRL1441
The G-401 [G401] is epithelial-like and isolated from the kidney of a White, 3-month, male with a rhabdoid tumor. This cell line was deposited by PT Peebles.