Learn More
Grainger Niroflex USA Niroflex2000 Chainmail Cut-Resistant Glove, Stainless Steel, Ambidextrous, Wrist Length, 1 ea
Catalog No. 502777012
Encompass_Preferred
These chainmail cut-resistant gloves provide limited protection from knife cuts and similar hazards, making them ideal for food processing, meat and poultry handling, and culinary applications. Constructed from high-quality stainless steel mesh, these five-finger gloves are designed for comfort, hygiene, and durability in professional food service environments.
  • Fits either right or left hand for ambidextrous use
  • Patented, easily adjustable wrist fastener for a secure fit
  • Features 1x4 interlocking pattern where each ring is interlocked with at least 4 adjacent rings
  • Clean quickly and easily without risk of contamination
  • Stainless steel construction with no fabric straps
  • Bar-coded serial number tag for tracking and scheduling
  • USDA approved for food processing applications
  • European specification EN1082-1 compliant
Catalog No. 50-277-7012 Supplier Grainger Supplier No. 18C897
These chainmail cut-resistant gloves provide limited protection from knife cuts and similar hazards, making them ideal for food processing, meat and poultry handling, and culinary applications. Constructed from high-quality stainless steel mesh, these five-finger gloves are designed for comfort, hygiene, and durability in professional food service environments.
  • Fits either right or left hand for ambidextrous use
  • Patented, easily adjustable wrist fastener for a secure fit
  • Features 1x4 interlocking pattern where each ring is interlocked with at least 4 adjacent rings
  • Clean quickly and easily without risk of contamination
  • Stainless steel construction with no fabric straps
  • Bar-coded serial number tag for tracking and scheduling
  • USDA approved for food processing applications
  • European specification EN1082-1 compliant