Learn More
Grainger Niroflex2000® Cut-Resistant Glove, Stainless Steel Mesh, 2XL/11, 1 ea
Catalog No. 502774422
Encompass_Preferred
These stainless steel chainmail gloves offer superior cut resistance for high-risk environments such as food processing, meat handling, and blade sharpening. Constructed from AISI 304L stainless steel, this metal mesh design ensures maximum durability and protection while maintaining flexibility for extended wear. The gloves feature a secure stainless steel clip closure system, facilitating easy cleaning and hygiene maintenance without the risk of fabric-related contamination.
  • Features 4:1 ring construction to minimize gaps and ensure high seam strength
  • Designed with a 3 in. extended cuff for added wrist protection
  • Includes metal-detectable, color-coded tags for efficient inventory management and compliance
  • Fully ambidextrous design for versatile use
  • Resistant to harsh cleaning chemicals and steam cleaning
  • Constructed from corrosion-resistant AISI 304L stainless steel
Catalog No. 50-277-4422 Supplier Grainger Supplier No. 18C910
These stainless steel chainmail gloves offer superior cut resistance for high-risk environments such as food processing, meat handling, and blade sharpening. Constructed from AISI 304L stainless steel, this metal mesh design ensures maximum durability and protection while maintaining flexibility for extended wear. The gloves feature a secure stainless steel clip closure system, facilitating easy cleaning and hygiene maintenance without the risk of fabric-related contamination.
  • Features 4:1 ring construction to minimize gaps and ensure high seam strength
  • Designed with a 3 in. extended cuff for added wrist protection
  • Includes metal-detectable, color-coded tags for efficient inventory management and compliance
  • Fully ambidextrous design for versatile use
  • Resistant to harsh cleaning chemicals and steam cleaning
  • Constructed from corrosion-resistant AISI 304L stainless steel