Learn More
Grainger Niroflex Niroflex2000 Metal Mesh Glove, Stainless Steel, Ambidextrous, XS/6, 1 ea
Catalog No. 502774406
Encompass_Preferred
These chainmail cut-resistant gloves are constructed from 100% stainless steel to provide a hygienic, durable solution for food processing environments. Designed to offer limited protection from knife cuts and similar hazards, the 5-finger, reversible design ensures versatility for either hand. Each ring is individually resistance welded to an average strength of 20 kg (0.20 kN) in a 1x4 interlocking pattern.
  • Manufactured in an ISO certified facility according to EN1082-1 specifications.
  • Features an easily adjustable, patented stainless steel wrist fastener.
  • Includes a bar-coded serial number tag for employee tracking and sanitation scheduling.
  • Equipped with a 3 in. extended thermoplastic polyurethane color-coded cuff.
  • Provides 5-finger coverage with a reversible design.
  • Accepted by the USDA Meat and Poultry Equipment Review Program.
Catalog No. 50-277-4406 Supplier Grainger Supplier No. 18C905
These chainmail cut-resistant gloves are constructed from 100% stainless steel to provide a hygienic, durable solution for food processing environments. Designed to offer limited protection from knife cuts and similar hazards, the 5-finger, reversible design ensures versatility for either hand. Each ring is individually resistance welded to an average strength of 20 kg (0.20 kN) in a 1x4 interlocking pattern.
  • Manufactured in an ISO certified facility according to EN1082-1 specifications.
  • Features an easily adjustable, patented stainless steel wrist fastener.
  • Includes a bar-coded serial number tag for employee tracking and sanitation scheduling.
  • Equipped with a 3 in. extended thermoplastic polyurethane color-coded cuff.
  • Provides 5-finger coverage with a reversible design.
  • Accepted by the USDA Meat and Poultry Equipment Review Program.