Learn More
Grainger Niroflex USA Niroflex2000 Chainmail Glove, Stainless Steel, Forearm, XL, 1 ea
Catalog No. 502774137
Encompass_Preferred
These stainless steel chainmail gloves provide limited protection from knife cuts and similar hazards in food processing applications, such as cutting meat or frozen food. Designed for a comfortable and secure fit, the gloves are often worn under disposable gloves to maintain hygiene standards.
  • 5-finger design provides comprehensive hand and forearm coverage
  • Ambidextrous construction fits either the right or left hand
  • 15.2 cm (6 in.) cuff features a thermoplastic polyurethane stiffener
  • Color-coded cuff identifies size for organization
  • Interlocking ring pattern includes at least 4 adjacent rings with a welding strength of 20 kg (0.20 kN) per ring
  • Easy to clean and sanitize without contamination risk
  • Bar-coded serial number tag facilitates employee tracking
  • Manufactured in an ISO certified facility and meets EN 1082-1 specifications
Catalog No. 50-277-4137 Supplier Grainger Supplier No. 18C916
These stainless steel chainmail gloves provide limited protection from knife cuts and similar hazards in food processing applications, such as cutting meat or frozen food. Designed for a comfortable and secure fit, the gloves are often worn under disposable gloves to maintain hygiene standards.
  • 5-finger design provides comprehensive hand and forearm coverage
  • Ambidextrous construction fits either the right or left hand
  • 15.2 cm (6 in.) cuff features a thermoplastic polyurethane stiffener
  • Color-coded cuff identifies size for organization
  • Interlocking ring pattern includes at least 4 adjacent rings with a welding strength of 20 kg (0.20 kN) per ring
  • Easy to clean and sanitize without contamination risk
  • Bar-coded serial number tag facilitates employee tracking
  • Manufactured in an ISO certified facility and meets EN 1082-1 specifications