Learn More
Invivogen G418 1G LIQUID @ 100MG/ML
SDP
Catalog No. NC9107150
Encompass
G418, 1g Liquid @ 100mg/mL, 5 x 2mL Tubes, Colorless Solution, Shipped at Room Temperature, Stored at -20degreeC, 1 Year, Aminoglycoside Antibiotic, Blocks Polypeptide Synthesis, by Inhibiting Elongation, Both Prokaryotic and Eukaryotic C
Catalog No. NC9107150 Supplier Invivogen Supplier No. ANTGN1
G418, 1g Liquid @ 100mg/mL, 5 x 2mL Tubes, Colorless Solution, Shipped at Room Temperature, Stored at -20degreeC, 1 Year, Aminoglycoside Antibiotic, Blocks Polypeptide Synthesis, by Inhibiting Elongation, Both Prokaryotic and Eukaryotic C