Learn More
TOKU-E G418 Disulfate (Geneticin Disulfate), 108321-42-2, 1 g
Catalog No. 502551800
Encompass_Preferred
G418 Disulfate (Geneticin Sulfate), aminoglycoside, selection antibiotic, C20H40N4O10 2H2SO4. Useful in cell culture, transfection.
Catalog No. 50-255-1800 Supplier TOKU-E Supplier No. G00125G
G418 Disulfate (Geneticin Sulfate), aminoglycoside, selection antibiotic, C20H40N4O10 2H2SO4. Useful in cell culture, transfection.