Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ G418 Disulfate PhytoPure™

Catalog No. 0215878283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-158-78283 250 mg
02-158-78291 1 g
02-158-78294 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-158-78283 Supplier MP Biomedicals™ Supplier No. 0215878283
Add to Cart
Add to Cart

G418 Disulfate PhytoPure™ is an aminoglycoside related to gentamicin that acts as an antibiotic and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths.

  • G418 Disulfate blocks polypeptide synthesis in both prokaryotic and eukaryotic cells and is useful for selection of genetically engineered eukarytoic cells in culture.
  • It is an effective selecting agent utilized in killing prokaryotic and eukaryotic cells.
  • It is also used for selection and maintenance of prokaryotic and eukaryotic cells transfected with a iNOS promoter construct and neomycin resistance gene.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108321-42-2
Color White
Content And Storage 2 to 8°C
Form Powder
Product Type Antibiotic
Sterility Non-sterile
Purity or Quality Grade Cell Culture Grade
Quantity 250 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.