Learn More
Enzo Life Sciences G418. sulfate (1g). CAS: 108321-42-2
SDP
Catalog No. 502008907
Encompass_Preferred
Gentamycin-class aminoglycoside antibiotic. Cytotoxic to prokaryotic and eukaryotic cells. Induces apoptosis. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3’ or APH II. Alternative name: Geneticin. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8907 Supplier Enzo Life Sciences Supplier No. ALX380013G001
Gentamycin-class aminoglycoside antibiotic. Cytotoxic to prokaryotic and eukaryotic cells. Induces apoptosis. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3’ or APH II. Alternative name: Geneticin. Solubility: Soluble in water. Long Term Storage: -20°C.