Learn More
AdipoGen G418 . sulfate (250 mg)
SDP
Supplier:  AdipoGen AGCN20030M250
Encompass_Preferred
Chemical. CAS: 108321-42-2. Formula: C20H40N4O10 . 2H2SO4. MW: 496.6 . 196.1. Isolated from Micromonospora rhodorangea. Antibiotic. Cytotoxic to prokaryotic and eukaryotic cells by inhibiting the protein synthesis. Apoptosis inducer. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3' or APH II. Antiviral.
Catalog No. 50-205-3375