Learn More
AdipoGen G418 . sulfate,Chemical. CAS: 108321-42-2
SDP
Catalog No. 50464353
Encompass_Preferred
G418 . sulfate, AdipoGen Life Sciences. Chemical. CAS: 108321-42-2. Formula: C20H40N4O10 . 2H2SO4. MW: 496.6 . 196.1. Isolated from Micromonospora rhodorangea. Antibiotic. Cytotoxic to prokaryotic and eukaryotic cells by inhibiting the protein synthesis. Apoptosis inducer. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3' or APH II. Antiviral.
Catalog No. 50-464-353 Supplier AdipoGen Supplier No. AGCN20030G005
G418 . sulfate, AdipoGen Life Sciences. Chemical. CAS: 108321-42-2. Formula: C20H40N4O10 . 2H2SO4. MW: 496.6 . 196.1. Isolated from Micromonospora rhodorangea. Antibiotic. Cytotoxic to prokaryotic and eukaryotic cells by inhibiting the protein synthesis. Apoptosis inducer. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3' or APH II. Antiviral.