Learn More
Enzo Life Sciences G418 . sulfate, (5g), CAS Number: 108321-42-2
SDP
Catalog No. NC9132228
Encompass_Preferred
Antibiotic. Gentamycin-class aminoglycoside antibiotic. Cytotoxic to prokaryotic and eukaryotic cells. Induces apoptosis. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3’ or APH II. Alternative Name: Geneticin. Formula: C20H40N4O10. 2H2SO4. MW: 496.6. 196.1. CAS Number: 108321-42-2. Appearance: White to off-white powder. Solubility: Soluble in water. Use/Stability: Solutions are stable for 3 to 6 months when stored at -20°C. Long Term Storage: -20°C
Catalog No. NC9132228 Supplier Enzo Life Sciences Supplier No. ALX380013G005
Antibiotic. Gentamycin-class aminoglycoside antibiotic. Cytotoxic to prokaryotic and eukaryotic cells. Induces apoptosis. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3’ or APH II. Alternative Name: Geneticin. Formula: C20H40N4O10. 2H2SO4. MW: 496.6. 196.1. CAS Number: 108321-42-2. Appearance: White to off-white powder. Solubility: Soluble in water. Use/Stability: Solutions are stable for 3 to 6 months when stored at -20°C. Long Term Storage: -20°C