Learn More
Electron Microscopy Sciences Zeiss Integrating Eyepiece Disc 100 Points G47, 21mm
SDP
Catalog No. 5029808
Encompass_Preferred
Similar to G49 but extended to 100 points which are indexed. Zeiss integrating eyepiece disc. Glass sandwich.
Size: 21 mm
Catalog No. 50-298-08 Supplier Electron Microscopy Sciences Supplier No. 6809421
Similar to G49 but extended to 100 points which are indexed. Zeiss integrating eyepiece disc. Glass sandwich.
Size: 21 mm