Learn More
Electron Microscopy Sciences Zeiss Integrating Eyepiece Disc 100 Points, 16mm
SDP
Catalog No. 5029810
Encompass_Preferred
Similar to G49 but extended to 100 points which are indexed. Zeiss integrating eyepiece disc. Glass sandwich.
Size: 16 mm
Catalog No. 50-298-10 Supplier Electron Microscopy Sciences Supplier No. 6809416
Similar to G49 but extended to 100 points which are indexed. Zeiss integrating eyepiece disc. Glass sandwich.
Size: 16 mm