Learn More
Electron Microscopy Sciences Henning Resseau Pattern Graticle G49, 16mm
SDP
Supplier:  Electron Microscopy Sciences 6809316
Encompass_Preferred
Zeiss Integrating Disc or Henning Reseau Pattern eyepiece reticle has 25 points on 6 horizontal lines.
Size: 16 mm
Catalog No. 50-298-07