Learn More
Electron Microscopy Sciences Henning Resseau Pattern Graticle G49, 21mm
SDP
Supplier:  Electron Microscopy Sciences 6809321
Encompass_Preferred
Zeiss Integrating Disc or Henning Reseau Pattern eyepiece reticle has 25 points on 6 horizontal lines.
Size: 21 mm
Catalog No. 50-298-11