Learn More
Medchemexpress LLC G4RGDSP, integrin-binding peptide | 774577-43-4 | 99.6% | 758.74 | 10 MG
SDP
Catalog No. 5000396157
Encompass_Preferred
G4RGDSP, integrin-binding peptide, is a cell integrin-binding peptide that targets integrin receptors. It can be coupled to alginate to increase the viability of cells in the scaffold. It is also used as an extrudable carrier for chondrocyte delivery for the study of 3D printing technology.
  • Targets integrin receptors
  • Can be coupled to alginate to increase cell viability in scaffolds
  • Used as an extrudable carrier for chondrocyte delivery for 3D printing technology studies
  • Increases expression of aggregin, collagen type II, and sox9 genes in HAC cells
Catalog No. 50-003-96157 Supplier Medchemexpress LLC Supplier No. HYP602210MG
G4RGDSP, integrin-binding peptide, is a cell integrin-binding peptide that targets integrin receptors. It can be coupled to alginate to increase the viability of cells in the scaffold. It is also used as an extrudable carrier for chondrocyte delivery for the study of 3D printing technology.
  • Targets integrin receptors
  • Can be coupled to alginate to increase cell viability in scaffolds
  • Used as an extrudable carrier for chondrocyte delivery for 3D printing technology studies
  • Increases expression of aggregin, collagen type II, and sox9 genes in HAC cells