Learn More
Medchemexpress LLC G4RGDSP, integrin-binding peptide | 774577-43-4 | 99.6% | 758.74 | 25 MG
SDP
Catalog No. 5000400328
Encompass_Preferred
G4RGDSP, Integrin-binding peptide is a cell integrin-binding peptide that targets integrin receptors. It is coupled to alginate to increase the viability of cells in the scaffold. It can be used as an extrudable carrier for chondrocyte delivery for the study of 3D printing technology.
  • Targets integrin receptors.
  • Can be coupled to alginate to increase cell viability in scaffolds.
  • Usable as an extrudable carrier for chondrocyte delivery for 3D printing technology.
  • Can increase the expression of aggregin, collagen type II, and sox9 genes in HAC cells.
Catalog No. 50-004-00328 Supplier Medchemexpress LLC Supplier No. 25MGHYP6022
G4RGDSP, Integrin-binding peptide is a cell integrin-binding peptide that targets integrin receptors. It is coupled to alginate to increase the viability of cells in the scaffold. It can be used as an extrudable carrier for chondrocyte delivery for the study of 3D printing technology.
  • Targets integrin receptors.
  • Can be coupled to alginate to increase cell viability in scaffolds.
  • Usable as an extrudable carrier for chondrocyte delivery for 3D printing technology.
  • Can increase the expression of aggregin, collagen type II, and sox9 genes in HAC cells.