Learn More
Medchemexpress LLC G4RGDSP, integrin-binding peptide | 774577-43-4 | 99.6% | 758.74 | 5 MG
SDP
Catalog No. 5000397832
Encompass_Preferred
G4RGDSP, Integrin-binding peptide, is a cell integrin-binding peptide designed to target integrin receptors. It is used in research and can be coupled to alginate to enhance cell viability in scaffolds. This peptide is also utilized as an extrudable carrier for chondrocyte delivery, particularly in studies involving 3D printing technology.
  • Cell integrin-binding peptide
  • Targets integrin receptors
  • Can be coupled to alginate to increase cell viability in scaffolds
  • Used as an extrudable carrier for chondrocyte delivery
  • Increases expression of aggregin, collagen type II, and sox9 genes in HAC cells
Catalog No. 50-003-97832 Supplier Medchemexpress LLC Supplier No. HYP60225MG
G4RGDSP, Integrin-binding peptide, is a cell integrin-binding peptide designed to target integrin receptors. It is used in research and can be coupled to alginate to enhance cell viability in scaffolds. This peptide is also utilized as an extrudable carrier for chondrocyte delivery, particularly in studies involving 3D printing technology.
  • Cell integrin-binding peptide
  • Targets integrin receptors
  • Can be coupled to alginate to increase cell viability in scaffolds
  • Used as an extrudable carrier for chondrocyte delivery
  • Increases expression of aggregin, collagen type II, and sox9 genes in HAC cells