Learn More
Medchemexpress LLC G4RGDSP, integrin-binding peptide | 774577-43-4 | 99.6% | 758.74 | 50 MG
SDP
Catalog No. 5000396158
Encompass_Preferred
G4RGDSP, integrin-binding peptide, is a cell integrin-binding peptide that specifically targets integrin receptors. It is designed to be coupled with alginate to enhance the viability of cells within a scaffold. This peptide can also be utilized as an extrudable carrier for chondrocyte delivery, facilitating studies involving 3D printing technology.
  • Targets integrin receptors as a cell integrin-binding peptide.
  • Can be coupled to alginate to increase cell viability in scaffolds.
  • Applicable as an extrudable carrier for chondrocyte delivery in 3D printing technology research.
  • Increases the expression of aggregin, collagen type II, and sox9 genes in HAC cells in vitro.
Catalog No. 50-003-96158 Supplier Medchemexpress LLC Supplier No. HYP602250MG
G4RGDSP, integrin-binding peptide, is a cell integrin-binding peptide that specifically targets integrin receptors. It is designed to be coupled with alginate to enhance the viability of cells within a scaffold. This peptide can also be utilized as an extrudable carrier for chondrocyte delivery, facilitating studies involving 3D printing technology.
  • Targets integrin receptors as a cell integrin-binding peptide.
  • Can be coupled to alginate to increase cell viability in scaffolds.
  • Applicable as an extrudable carrier for chondrocyte delivery in 3D printing technology research.
  • Increases the expression of aggregin, collagen type II, and sox9 genes in HAC cells in vitro.