Learn More
Medchemexpress LLC G4RGDSP, integrin-binding peptide | 774577-43-4 | 99.61% | 758.74 | 100 MG
SDP
Catalog No. 5000397109
Encompass_Preferred
G4RGDSP, integrin-binding peptide is a cell integrin-binding peptide that targets integrin receptors. It is coupled to alginate to increase the viability of cells in the scaffold and can be used as an extrudable carrier for chondrocyte delivery for the study of 3D printing technology.
  • Targets integrin receptors
  • Increases viability of cells in scaffold
  • Suitable for chondrocyte delivery studies
  • Used in 3D printing technology research
  • Increases expression of aggregin, collagen type II, and sox9 genes in HAC cells
Catalog No. 50-003-97109 Supplier Medchemexpress LLC Supplier No. HYP6022100MG
G4RGDSP, integrin-binding peptide is a cell integrin-binding peptide that targets integrin receptors. It is coupled to alginate to increase the viability of cells in the scaffold and can be used as an extrudable carrier for chondrocyte delivery for the study of 3D printing technology.
  • Targets integrin receptors
  • Increases viability of cells in scaffold
  • Suitable for chondrocyte delivery studies
  • Used in 3D printing technology research
  • Increases expression of aggregin, collagen type II, and sox9 genes in HAC cells