Learn More
Medchemexpress LLC G5-7 10mg | 939681-36-4 | 383.39 g/mol | C22H19F2NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11545210MG
Encompass_Preferred
G5-7 is an orally active, allosteric JAK2 inhibitor intended for research use. It selectively inhibits JAK2-mediated phosphorylation and activation of EGFR (Tyr1068) and STAT3, and has been reported to induce cell cycle arrest, apoptosis, and exhibit antiangiogenic effects relevant to glioma studies.
  • Allosteric JAK2 inhibitor with selective inhibition of EGFR (Tyr1068) and STAT3 phosphorylation.
  • Reported to induce cell cycle arrest and apoptosis in cellular studies.
  • Exhibits antiangiogenic activity relevant to glioma research.
  • Off-white to light yellow solid powder suitable for research use.
  • High purity (99.55%) to support experimental reproducibility.
  • Recommended storage: powder at -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-08006