Learn More
Medchemexpress LLC G5-7 25mg | 939681-36-4 | 25 MG
SDP
Catalog No. 5000308005
Encompass_Preferred
G5-7 is an orally active, allosteric JAK2 inhibitor used for research applications. It selectively inhibits JAK2-mediated phosphorylation and activation of EGFR (Tyr1068) and STAT3, and has been reported to induce cell cycle arrest, promote apoptosis, and exhibit antiangiogenic effects.
  • Allosteric JAK2 inhibitor with oral activity.
  • Selective inhibition of JAK2-mediated EGFR and STAT3 phosphorylation.
  • Reported to induce cell cycle arrest and apoptosis.
  • Exhibits antiangiogenic activity and glioma research relevance.
  • Provided as a solid; 25 mg pack available.
  • High purity (approximately 99.6%).
Catalog No. 50-003-08005 Supplier Medchemexpress LLC Supplier No. HY11545225MG
G5-7 is an orally active, allosteric JAK2 inhibitor used for research applications. It selectively inhibits JAK2-mediated phosphorylation and activation of EGFR (Tyr1068) and STAT3, and has been reported to induce cell cycle arrest, promote apoptosis, and exhibit antiangiogenic effects.
  • Allosteric JAK2 inhibitor with oral activity.
  • Selective inhibition of JAK2-mediated EGFR and STAT3 phosphorylation.
  • Reported to induce cell cycle arrest and apoptosis.
  • Exhibits antiangiogenic activity and glioma research relevance.
  • Provided as a solid; 25 mg pack available.
  • High purity (approximately 99.6%).