Learn More
Medchemexpress LLC HY-115452 5mg Medchemexpress, G5-7 CAS:939681-36-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1154525MG
Encompass_Preferred
Medchemexpress, HY-115452 5mg G5-7 CAS:939681-36-4 G5-7, an orally active and allosteric JAK2 inhibitor, selectively inhibits JAK2 mediated phosphorylation and activation of EGFR (Tyr1068) and STAT3 by binding to JAK2. G5-7 induces cell cycle arrest, apoptosis and possesses antiangiogenic effect. G5-7 has the potential for glioma study. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0879