Learn More
Epoch Life Science Inc EconoSpin® prepacked G50 Spin Columns with inert support for Dye Terminator Removal, Desalting, Pre-filtration etc. With screw cap and snap off end-tail (250 columns)
SDP
Catalog No. 501538110
Encompass_Preferred
PP housing with prepacked G50 on an inert support. 700 ul bed volume. 50-200 ul sample volume. Liquid tight screw cap enables long term buffer storage and eliminates splash when opening/closing. Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 250 columns.
Catalog No. 50-153-8110 Supplier Epoch Life Science Inc Supplier No. 2080250
PP housing with prepacked G50 on an inert support. 700 ul bed volume. 50-200 ul sample volume. Liquid tight screw cap enables long term buffer storage and eliminates splash when opening/closing. Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 250 columns.