Learn More
Grainger G5368 ProtCase 4 7/64 in PwrClwLtcSys/PdLk Or
Catalog No. 502767916
Encompass_Preferred
The Proto Crowfoot Socket Wrench is a high quality alloy steel crowfoot wrench designed for use in tight spaces. It has a 15 degree offset head with a 1/2 inch square drive to allow access where a straight head wrench won't fit. The wrench comes in multiple sizes from 5/16 inch to 1 inch and is made in the USA. This durable crowfoot wrench can deliver up to 75 ft-lbs of torque.
Catalog No. 50-276-7916 Supplier Grainger Supplier No. 16Z224
The Proto Crowfoot Socket Wrench is a high quality alloy steel crowfoot wrench designed for use in tight spaces. It has a 15 degree offset head with a 1/2 inch square drive to allow access where a straight head wrench won't fit. The wrench comes in multiple sizes from 5/16 inch to 1 inch and is made in the USA. This durable crowfoot wrench can deliver up to 75 ft-lbs of torque.