Learn More
Grainger G5370 ProtCase 4 7/64 in PwrClwLtcSys/PdLk Or
Catalog No. 502768189
Encompass_Preferred
The Grainger 16Z301 is a 3M Novec 1230 Clean Agent Fire Extinguisher, 2.5 lb capacity, UL rated 2-B:C, features a steel cylinder with red powder coat finish, a commercial valve assembly with pressure gauge, and a wall hook. It uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable electronics and equipment. This compact and lightweight fire extinguisher is easy to handle and can suppress Class A, B, and C fires. With proper care and maintenance, it provides reliable fire protection.
Catalog No. 50-276-8189 Supplier Grainger Supplier No. 16Z301
The Grainger 16Z301 is a 3M Novec 1230 Clean Agent Fire Extinguisher, 2.5 lb capacity, UL rated 2-B:C, features a steel cylinder with red powder coat finish, a commercial valve assembly with pressure gauge, and a wall hook. It uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable electronics and equipment. This compact and lightweight fire extinguisher is easy to handle and can suppress Class A, B, and C fires. With proper care and maintenance, it provides reliable fire protection.