Learn More
Grainger G5370 ProtCase 4 7/64 in PwrClwLtcSys/PdLk Ylw
Catalog No. 502767803
Encompass_Preferred
The Grainger Approved G5370 ProtCase is a rugged, waterproof case designed to protect electronic devices. It features an IP67 waterproof rating, meaning it is dust tight and can be immersed in 1 meter of water for 30 minutes. The case is made from durable polycarbonate and has a rubber seal to keep out moisture. It has an adjustable padded interior to secure items in place. The case has a clear back so device screens can still be used while inside. It comes with a detachable belt clip for easy transport. The case dimensions are 5.7" x 3.2" x 1.5" and it weighs 5.6 oz.
Catalog No. 50-276-7803 Supplier Grainger Supplier No. 16Z302
The Grainger Approved G5370 ProtCase is a rugged, waterproof case designed to protect electronic devices. It features an IP67 waterproof rating, meaning it is dust tight and can be immersed in 1 meter of water for 30 minutes. The case is made from durable polycarbonate and has a rubber seal to keep out moisture. It has an adjustable padded interior to secure items in place. The case has a clear back so device screens can still be used while inside. It comes with a detachable belt clip for easy transport. The case dimensions are 5.7" x 3.2" x 1.5" and it weighs 5.6 oz.